3-(Aminomethyl)-1-tosylpyrrole

Lieferant: Apollo Scientific

APOSOR471260-1GEA 1540 EUR
APOSOR471260-1G
3-(Aminomethyl)-1-tosylpyrrole
3-(Aminomethyl)-1-tosylpyrrole
Formel: C₁₂H₁₄N₂O₂S
Molecular Weight: 250,317 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28398151
CAS-Nummer: 872405-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR