Methyl (E)-3-Morpholinoacrylate

Lieferant: Apollo Scientific

APOSOR471149-1GEA 499 EUR
APOSOR471149-1G
Methyl (E)-3-Morpholinoacrylate
Methyl (E)-3-Morpholinoacrylate
Formel: C₈H₁₃NO₃
Molecular Weight: 171,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01696261
CAS-Nummer: 101471-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR