Methyl 1-Benzylimidazole-2-carboxylate 95%

Lieferant: Apollo Scientific

APOSOR471144-250MGEA 292 EUR
APOSOR471144-250MG
Methyl 1-Benzylimidazole-2-carboxylate 95%
Methyl 1-Benzylimidazole-2-carboxylate
Formel: C₁₂H₁₂N₂O₂
Molecular Weight: 216,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26680731
CAS-Nummer: 1502811-19-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR