Ethyl 3-(2-Cyano-N-methylacetamido)propanoate

Lieferant: Apollo Scientific

APOSOR471079-1GEA 1120 EUR
APOSOR471079-1G
Ethyl 3-(2-Cyano-N-methylacetamido)propanoate
Ethyl 3-(2-Cyano-N-methylacetamido)propanoate
Formel: C₉H₁₄N₂O₃
Molecular Weight: 198,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16147409
CAS-Nummer: 1306948-45-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR