4,6-Dihydroxycoumarin
Lieferant: Apollo Scientific
APOSOR471074-1GEA
713
EUR
APOSOR471074-1G
4,6-Dihydroxycoumarin
4,6-Dihydroxycoumarin
Formel:
C₉H₆O₄ Molecular Weight: 178,142 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00464231 CAS-Nummer: 30992-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...