Methyl 2-Phenylisonicotinate

Lieferant: Apollo Scientific

APOSOR471071-5GEA 1120 EUR
APOSOR471071-5G
Methyl 2-Phenylisonicotinate
Methyl 2-Phenylisonicotinate
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07357447
CAS-Nummer: 4634-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR