N-[(3-Pyridyl)methyl]-2-propanamine
Lieferant: Apollo Scientific
APOSOR470956-5GEA
529
EUR
APOSOR470956-5G
N-[(3-Pyridyl)methyl]-2-propanamine
N-[(3-Pyridyl)methyl]-2-propanamine
Formel:
C₉H₁₄N₂ Molecular Weight: 150,221 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00716868 CAS-Nummer: 19730-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...