Ethyl 5-Bromoisoquinoline-1-carboxylate

Lieferant: Apollo Scientific

APOSOR470933-1GEA 1150 EUR
APOSOR470933-1G
Ethyl 5-Bromoisoquinoline-1-carboxylate
Ethyl 5-Bromoisoquinoline-1-carboxylate
Formel: C₁₂H₁₀BrNO₂
Molecular Weight: 280,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27986819
CAS-Nummer: 1111311-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR