5-Bromo-N,N-diethyl-1-pentanamine hydrobromide
Lieferant: Apollo Scientific
APOSOR470746-1GEA
1120
EUR
APOSOR470746-1G
5-Bromo-N,N-diethyl-1-pentanamine hydrobromide
5-Bromo-N,N-diethyl-1-pentanamine hydrobromide
Formel:
C₉H₂₀BrN·BrH Molecular Weight: 303,078 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27500643 CAS-Nummer: 2006277-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...