3-Amino-5-bromo-4-methoxypyridine
Lieferant: Apollo Scientific
APOSOR470624-1GEA
804
EUR
APOSOR470624-1G
3-Amino-5-bromo-4-methoxypyridine
3-Amino-5-bromo-4-methoxypyridine
Formel:
C₆H₇BrN₂O Molecular Weight: 203,037 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100691 CAS-Nummer: 31872-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...