1-Phenyl-1-(3-quinolyl)ethanol

Lieferant: Apollo Scientific

APOSOR470550-1GEA 601 EUR
APOSOR470550-1G
1-Phenyl-1-(3-quinolyl)ethanol
1-Phenyl-1-(3-quinolyl)ethanol
Formel: C₁₇H₁₅NO
Molecular Weight: 249,307 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12833780
CAS-Nummer: 1183075-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR