Ethyl 3-Bromo-4-chloroindole-1-carboxylate

Lieferant: Apollo Scientific

APOSOR470496-5GEA 713 EUR
APOSOR470496-5G
Ethyl 3-Bromo-4-chloroindole-1-carboxylate
Ethyl 3-Bromo-4-chloroindole-1-carboxylate
Formel: C₁₁H₉BrClNO₂
Molecular Weight: 302,552 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22205874
CAS-Nummer: 1375064-58-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR