(8-Phenyl-1,4-dioxaspiro[4.5]dec-8-yl)methanamine

Lieferant: Apollo Scientific

APOSOR470282-5GEA 2020 EUR
APOSOR470282-5G
(8-Phenyl-1,4-dioxaspiro[4.5]dec-8-yl)methanamine
(8-Phenyl-1,4-dioxaspiro[4.5]dec-8-yl)methanamine
Formel: C₁₅H₂₁NO₂
Molecular Weight: 247,333 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410580
CAS-Nummer: 443687-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR