2-Cyano-N-cyclobutyl-acetamide
Lieferant: Apollo Scientific
APOSOR451170-1GEA
684
EUR
APOSOR451170-1G
2-Cyano-N-cyclobutyl-acetamide
2-Cyano-N-cyclobutyl-acetamide
Formel:
C₇H₁₀N₂O Molecular Weight: 138,167 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16755845 CAS-Nummer: 1064448-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...