4-(Tetrahydro-2H-pyran-4-yloxy)benzonitrile

Lieferant: Apollo Scientific

APOSOR42103-5GEA 487 EUR
APOSOR42103-5G APOSOR42103-1G
4-(Tetrahydro-2H-pyran-4-yloxy)benzonitrile
4-(Tetrahydro-2H-pyran-4-yloxy)benzonitrile
Formel: C₁₂H₁₃NO₂
Molecular Weight: 203,237 g/mol
Schmelzpunkt: 109…111,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025875
CAS-Nummer: 884507-34-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR