1-(2-Pentyl)piperazine

Achtung

APOSOR4169-1GEA 46.6 EUR
APOSOR4169-1G APOSOR4169-5G APOSOR4169-10G
1-(2-Pentyl)piperazine
1-(2-Pentyl)piperazine
Formel: C₉H₂₀N₂
Molecular Weight: 156,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093553
CAS-Nummer: 82499-96-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR