3-(Hydroxymethyl)cyclohexan-1-one 90%
Lieferant: Apollo Scientific
|
Achtung
|
APOSOR40418-1GEA
803
EUR
APOSOR40418-1G
3-(Hydroxymethyl)cyclohexan-1-one 90%
3-(Hydroxymethyl)cyclohexan-1-one
Formel:
C₇H₁₂O₂ Molecular Weight: 128,169 g/mol Siedepunkt: 135…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09414734 CAS-Nummer: 32916-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...