5-Bromo-2-(dimethylamino)benzaldehyde

Lieferant: Apollo Scientific

APOSOR400566-1GEA 205 EUR
APOSOR400566-1G
5-Bromo-2-(dimethylamino)benzaldehyde
5-Bromo-2-(dimethylamino)benzaldehyde
Formel: C₉H₁₀BrNO
Molecular Weight: 228,086 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20529529
CAS-Nummer: 171881-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR