2-Bromo-4-chlorobenzyl alcohol

Lieferant: Apollo Scientific

APOSOR400048-1GEA 364 EUR
APOSOR400048-1G APOSOR400048-5G
2-Bromo-4-chlorobenzyl alcohol
2-Bromo-4-chlorobenzyl alcohol
Formel: C₇H₆BrClO
Molecular Weight: 221,479 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09056765
CAS-Nummer: 143888-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR