2-(5-Bromo-2-methyl-2H-indazol-3-yl)propan-2-ol 99%

Lieferant: Apollo Scientific

APOSOR370107-250MGEA 1320 EUR
APOSOR370107-250MG
2-(5-Bromo-2-methyl-2H-indazol-3-yl)propan-2-ol 99%
2-(5-Bromo-2-methyl-2H-indazol-3-yl)propan-2-ol
Formel: C₁₁H₁₃BrN₂O
Molecular Weight: 269,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28359285
CAS-Nummer: 1823787-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR