4-Chloro-3H-Imidazo[4,5-c]quinoline, tech

Lieferant: Apollo Scientific

APOSOR370100-250MGEA 1900 EUR
APOSOR370100-250MG
4-Chloro-3H-Imidazo[4,5-c]quinoline, tech
4-Chloro-3H-Imidazo[4,5-c]quinoline
Formel: C₁₀H₆ClN₃
Molecular Weight: 203,628 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13151921
CAS-Nummer: 132206-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR