N,2,3-Trimethyl-2H-indazol-6-amine ≥98%

Lieferant: Apollo Scientific

APOSOR370084-500MGEA 1660 EUR
APOSOR370084-500MG
N,2,3-Trimethyl-2H-indazol-6-amine ≥98%
N,2,3-Trimethyl-2H-indazol-6-amine
Formel: C₁₀H₁₃N₃
Molecular Weight: 175,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24567305
CAS-Nummer: 1376676-65-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR