2-Chloro-3-ethoxyphenylboronic acid

Lieferant: Apollo Scientific
Achtung

APOSOR360929-1GEA 364 EUR
APOSOR360929-1G
2-Chloro-3-ethoxyphenylboronic acid
2-Chloro-3-ethoxyphenylboronic acid
Formel: C₈H₁₀BClO₃
Molecular Weight: 200,427 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17015726
CAS-Nummer: 1256345-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR