(R)-1,1,2-Triphenyl-2-(piperidin-1-yl)ethanol

Lieferant: Apollo Scientific

APOSOR350341-1GEA 473 EUR
APOSOR350341-1G
(R)-1,1,2-Triphenyl-2-(piperidin-1-yl)ethanol
(R)-1,1,2-Triphenyl-2-(piperidin-1-yl)ethanol
Formel: C₂₅H₂₇NO
Molecular Weight: 357,488 g/mol
Schmelzpunkt: 159…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03427065
CAS-Nummer: 213995-12-1
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR