4-Methoxy-2-vinylpyridine 95+%

Lieferant: Apollo Scientific

APOSOR346662-1GEA 622 EUR
APOSOR346662-1G
4-Methoxy-2-vinylpyridine 95+%
4-Methoxy-2-vinylpyridine
Formel: C₈H₉NO
Molecular Weight: 135,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384441
CAS-Nummer: 30566-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR