2-(Pyrid-2-yloxy)benzaldehyde 95+%

Lieferant: Apollo Scientific

APOSOR346637-1GEA 166 EUR
APOSOR346637-1G
2-(Pyrid-2-yloxy)benzaldehyde 95+%
2-(Pyrid-2-yloxy)benzaldehyde
Formel: C₁₂H₉NO₂
Molecular Weight: 199,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025852
CAS-Nummer: 141580-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR