Benzo[d]isoxazole-5-sulphonyl chloride

Gefahr

APOSOR346294-1GEA 0 EUR
APOSOR346294-1G
Benzo[d]isoxazole-5-sulphonyl chloride
Benzo[d]isoxazole-5-sulphonyl chloride
Formel: C₇H₄ClNO₃S
Molecular Weight: 217,63 g/mol
Schmelzpunkt: 77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659651
CAS-Nummer: 16331-62-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR