5-Bromo-4-chloro-6-methoxypyrimidine

Lieferant: Apollo Scientific
Achtung

APOSOR345715-250MGEA 331 EUR
APOSOR345715-250MG APOSOR345715-1G
5-Bromo-4-chloro-6-methoxypyrimidine
5-Bromo-4-chloro-6-methoxypyrimidine
Formel: C₅H₄BrClN₂O
Molecular Weight: 223,455 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234033
CAS-Nummer: 4319-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR