(4R)-1-Methyl-4-(prop-1-en-2-yl)-7-azabicyclo[4.1.0]heptane

Lieferant: Apollo Scientific

APOSOR340187-100MGEA 459 EUR
APOSOR340187-100MG
(4R)-1-Methyl-4-(prop-1-en-2-yl)-7-azabicyclo[4.1.0]heptane
(4R)-1-Methyl-4-(prop-1-en-2-yl)-7-azabicyclo[4.1.0]heptane
Formel: C₁₀H₁₇N
Molecular Weight: 151,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28348392
CAS-Nummer: 55854-40-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR