(3-(7-((tert-Butyldiphenylsilyl)oxy)heptyl)aziridin-2-yl)methanol

Lieferant: Apollo Scientific

APOSOR340176-100MGEA 706 EUR
APOSOR340176-100MG
(3-(7-((tert-Butyldiphenylsilyl)oxy)heptyl)aziridin-2-yl)methanol
(3-(7-((tert-Butyldiphenylsilyl)oxy)heptyl)aziridin-2-yl)methanol
Formel: C₂₆H₃₉NO₂Si
Molecular Weight: 425,679 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154780
CAS-Nummer: 1980035-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR