2-(o-Tolyl)benzofuran
APOSOR340159-100MGEA
0
EUR
APOSOR340159-100MG
2-(o-Tolyl)benzofuran
2-(o-Tolyl)benzofuran
Formel:
C₁₅H₁₂O Molecular Weight: 208,255 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28154770 CAS-Nummer: 65246-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...