Methyl 2-(3-cyano-4-nitrophenyl)-2-(o-tolyl)acetate

Lieferant: Apollo Scientific

APOSOR340143-100MGEA 1930 EUR
APOSOR340143-100MG
Methyl 2-(3-cyano-4-nitrophenyl)-2-(o-tolyl)acetate
Methyl 2-(3-cyano-4-nitrophenyl)-2-(o-tolyl)acetate
Formel: C₁₇H₁₄N₂O₄
Molecular Weight: 310,304 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154756
CAS-Nummer: 1848979-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR