8H-Thieno[2,3-b]indole

Lieferant: Apollo Scientific

APOSOR340104-100MGEA 626 EUR
APOSOR340104-100MG
8H-Thieno[2,3-b]indole
8H-Thieno[2,3-b]indole
Formel: C₁₀H₇NS
Molecular Weight: 173,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13178619
CAS-Nummer: 23989-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR