1-(6-(Methylsulphonamido)-3-methoxy-2,4-dimethylphenyl)-7-methoxynaphthalen-2-ol

Lieferant: Apollo Scientific

APOSOR340079-50MGEA 1760 EUR
APOSOR340079-50MG
1-(6-(Methylsulphonamido)-3-methoxy-2,4-dimethylphenyl)-7-methoxynaphthalen-2-ol
1-(6-(Methylsulphonamido)-3-methoxy-2,4-dimethylphenyl)-7-methoxynaphthalen-2-ol
Formel: C₂₁H₂₃NO₅S
Molecular Weight: 401,476 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154711
CAS-Nummer: 1832514-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR