2'-Amino-3'-chloro-4'-methyl-[1,1'-biphenyl]-2-ol

APOSOR340051-50MGEA 0 EUR
APOSOR340051-50MG
2'-Amino-3'-chloro-4'-methyl-[1,1'-biphenyl]-2-ol
2'-Amino-3'-chloro-4'-methyl-[1,1'-biphenyl]-2-ol
Formel: C₁₃H₁₂ClNO
Molecular Weight: 233,693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154684
CAS-Nummer: 1424386-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR