2-Chloro-4-(methoxymethyl)-6-methylquinolin-7-amine, tech

Lieferant: Apollo Scientific

APOSOR33173-1GEA 1390 EUR
APOSOR33173-1G
2-Chloro-4-(methoxymethyl)-6-methylquinolin-7-amine, tech
2-Chloro-4-(methoxymethyl)-6-methylquinolin-7-amine
Formel: C₁₂H₁₃ClN₂O
Molecular Weight: 236,697 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03848702
CAS-Nummer: 866134-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR