2-(2-Hydroxyethyl)-6-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech

Lieferant: Apollo Scientific

APOSOR33165-1GEA 1390 EUR
APOSOR33165-1G
2-(2-Hydroxyethyl)-6-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one, tech
2-(2-Hydroxyethyl)-6-methyl-3,4-dihydro-2H-1,4-benzoxazin-3-one
Formel: C₁₁H₁₃NO₃
Molecular Weight: 207,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03848651
CAS-Nummer: 191096-42-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR