4-Chloro-6-(4-methylpiperazin-1-yl)pyrimidin-2-amine hydrochloride, tech

Lieferant: Apollo Scientific

APOSOR33125-1GEA 1390 EUR
APOSOR33125-1G
4-Chloro-6-(4-methylpiperazin-1-yl)pyrimidin-2-amine hydrochloride, tech
4-Chloro-6-(4-methylpiperazin-1-yl)pyrimidin-2-amine hydrochloride
Formel: C₉H₁₄ClN₅·ClH
Molecular Weight: 264,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06795616
CAS-Nummer: 321571-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR