(2E)-1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)prop-2-en-1-one, tech

Lieferant: Apollo Scientific

APOSOR33124-1GEA 1390 EUR
APOSOR33124-1G
(2E)-1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)prop-2-en-1-one, tech
(2E)-1-(2,5-Dichlorothiophen-3-yl)-3-(dimethylamino)prop-2-en-1-one
Formel: C₉H₉Cl₂NOS
Molecular Weight: 250,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01443682
CAS-Nummer: 166196-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR