3-(4-Methoxyphenyl)-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione, tech
Lieferant: Apollo Scientific
APOSOR33114-1GEA
1390
EUR
APOSOR33114-1G
3-(4-Methoxyphenyl)-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione, tech
3-(4-Methoxyphenyl)-1-(phenylamino)-2,5-dihydro-1H-pyrrole-2,5-dione
Formel:
C₁₇H₁₄N₂O₃ Molecular Weight: 294,305 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00974747 CAS-Nummer: 49811-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...