4-Methoxy-2-phenoxypyridine-3-carbonitrile, tech

Lieferant: Apollo Scientific

APOSOR33112-1GEA 1390 EUR
APOSOR33112-1G
4-Methoxy-2-phenoxypyridine-3-carbonitrile, tech
4-Methoxy-2-phenoxypyridine-3-carbonitrile
Formel: C₁₃H₁₀N₂O₂
Molecular Weight: 226,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00975465
CAS-Nummer: 339016-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR