N'-[(1Z)-[4-(Dimethylamino)phenyl]methylidene]methoxycarbohydrazide, tech

Lieferant: Apollo Scientific

APOSOR33108-1GEA 1390 EUR
APOSOR33108-1G
N'-[(1Z)-[4-(Dimethylamino)phenyl]methylidene]methoxycarbohydrazide, tech
N'-[(1Z)-[4-(Dimethylamino)phenyl]methylidene]methoxycarbohydrazide
Formel: C₁₁H₁₅N₃O₂
Molecular Weight: 221,256 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00664684
CAS-Nummer: 339016-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR