3-((E)-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid 98%

Lieferant: Apollo Scientific

Achtung

APOSOR3310-1GEA 93.1 EUR
APOSOR3310-1G APOSOR3310-5G
3-((E)-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid 98%
3-((E)-3-Ethoxy-3-oxo-1-propen-1-yl)benzeneboronic acid
Formel: C₁₁H₁₃BO₄
Molecular Weight: 220,03 g/mol
Schmelzpunkt: 141…143 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08689480
CAS-Nummer: 913835-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR