(5E)-4-(4-Chlorophenyl)-5-[(dimethylamino)methylidene]-2-oxo-2,5-dihydrofuran-3-carbonitrile, tech

Lieferant: Apollo Scientific

APOSOR33062-1GEA 1390 EUR
APOSOR33062-1G
(5E)-4-(4-Chlorophenyl)-5-[(dimethylamino)methylidene]-2-oxo-2,5-dihydrofuran-3-carbonitrile, tech
(5E)-4-(4-Chlorophenyl)-5-[(dimethylamino)methylidene]-2-oxo-2,5-dihydrofuran-3-carbonitrile
Formel: C₁₄H₁₁ClN₂O₂
Molecular Weight: 274,702 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571828
CAS-Nummer: 50691-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR