N-Benzyl-1H-indole-3-carboxamide 95%

Lieferant: Apollo Scientific

APOSOR33035-1GEA 1070 EUR
APOSOR33035-1G
N-Benzyl-1H-indole-3-carboxamide 95%
N-Benzyl-1H-indole-3-carboxamide
Formel: C₁₆H₁₄N₂O
Molecular Weight: 250,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03305733
CAS-Nummer: 61788-25-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR