1-(1H-1,2,3-Benzotriazol-1-yl)propan-2-one 95%
Lieferant: Apollo Scientific
APOSOR33029-1GEA
640
EUR
APOSOR33029-1G
1-(1H-1,2,3-Benzotriazol-1-yl)propan-2-one 95%
1-(1H-1,2,3-Benzotriazol-1-yl)propan-2-one
Formel:
C₉H₉N₃O Molecular Weight: 175,187 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00825993 CAS-Nummer: 64882-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...