N'-(6-Chloropyridin-2-yl)cyclopropanecarbohydrazide, tech
Lieferant: Apollo Scientific
APOSOR33020-1GEA
1390
EUR
APOSOR33020-1G
N'-(6-Chloropyridin-2-yl)cyclopropanecarbohydrazide, tech
N'-(6-Chloropyridin-2-yl)cyclopropanecarbohydrazide
Formel:
C₉H₁₀ClN₃O Molecular Weight: 211,648 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02187333 CAS-Nummer: 478064-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...