3-Amino-5-(4-chlorophenyl)cyclohex-2-en-1-one, tech
Lieferant: Apollo Scientific
APOSOR33019-1GEA
1390
EUR
APOSOR33019-1G
3-Amino-5-(4-chlorophenyl)cyclohex-2-en-1-one, tech
3-Amino-5-(4-chlorophenyl)cyclohex-2-en-1-one
Formel:
C₁₂H₁₂ClNO Molecular Weight: 221,683 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00105289 CAS-Nummer: 478064-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...