Ethyl 4-(2-oxopyrrolidin-1-yl)benzoate, tech
Lieferant: Apollo Scientific
APOSOR32885-1GEA
1390
EUR
APOSOR32885-1G
Ethyl 4-(2-oxopyrrolidin-1-yl)benzoate, tech
Ethyl 4-(2-oxopyrrolidin-1-yl)benzoate
Formel:
C₁₃H₁₅NO₃ Molecular Weight: 233,263 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00973916 CAS-Nummer: 86364-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...