3-[2-(4-Chlorophenyl)hydrazin-1-ylidene]pentane-2,4-dione, tech
Lieferant: Apollo Scientific
APOSOR32877-1GEA
1390
EUR
APOSOR32877-1G
3-[2-(4-Chlorophenyl)hydrazin-1-ylidene]pentane-2,4-dione, tech
3-[2-(4-Chlorophenyl)hydrazin-1-ylidene]pentane-2,4-dione
Formel:
C₁₁H₁₁ClN₂O₂ Molecular Weight: 238,67 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00101089 CAS-Nummer: 24756-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...